Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35402297-35402552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFKBIA
Entrez Gene ID
4792
Gene Name
NFKB inhibitor alpha
Gene Aliases
IKBA, MAD-3, NFKBI
UniGene
Hs.81328
Gene Chromosome Location
Chr.14: 35401510-35404754 on Build GRCh38
Transcripts
NM_020529.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645840 c.866A>G p.Y289C Chr.14: 35402434-35402434 on Build GRCh38
6586281 c.872_873delCA p.T291fs*15 Chr.14: 35402427-35402428 on Build GRCh38
5713972 c.763C>T p.Q255* Chr.14: 35402537-35402537 on Build GRCh38
6976856 c.808G>A p.G270S Chr.14: 35402492-35402492 on Build GRCh38
6974581 c.802C>A p.Q268K Chr.14: 35402498-35402498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765954297 Chr.14: 35402354-35402354 on Build GRCh38
rs777020702 Chr.14: 35402406-35402406 on Build GRCh38
rs965141856 Chr.14: 35402381-35402381 on Build GRCh38
rs770202858 Chr.14: 35402529-35402529 on Build GRCh38
rs756020078 Chr.14: 35402459-35402459 on Build GRCh38
rs751355248 Chr.14: 35402357-35402357 on Build GRCh38
rs201191831 Chr.14: 35402413-35402413 on Build GRCh38
rs755221348 Chr.14: 35402515-35402515 on Build GRCh38
rs781464425 Chr.14: 35402521-35402521 on Build GRCh38
rs772607545 Chr.14: 35402424-35402424 on Build GRCh38
rs752524885 Chr.14: 35402457-35402457 on Build GRCh38
rs769191348 Chr.14: 35402404-35402404 on Build GRCh38
rs781022181 Chr.14: 35402370-35402370 on Build GRCh38
rs1033027351 Chr.14: 35402347-35402347 on Build GRCh38
rs774043011 Chr.14: 35402438-35402438 on Build GRCh38
rs749942106 Chr.14: 35402345-35402345 on Build GRCh38
rs957688002 Chr.14: 35402434-35402434 on Build GRCh38
rs765457814 Chr.14: 35402365-35402366 on Build GRCh38
rs146132589 Chr.14: 35402397-35402397 on Build GRCh38
rs1003119861 Chr.14: 35402325-35402325 on Build GRCh38
rs376370831 Chr.14: 35402517-35402517 on Build GRCh38
rs139180460 Chr.14: 35402412-35402412 on Build GRCh38
rs758502941 Chr.14: 35402499-35402499 on Build GRCh38
rs750562815 Chr.14: 35402478-35402478 on Build GRCh38
rs111375176 Chr.14: 35402393-35402393 on Build GRCh38
rs766046360 Chr.14: 35402428-35402428 on Build GRCh38
rs910465669 Chr.14: 35402422-35402422 on Build GRCh38
rs762648255 Chr.14: 35402346-35402346 on Build GRCh38
rs754156035 Chr.14: 35402360-35402363 on Build GRCh38
rs767285341 Chr.14: 35402456-35402456 on Build GRCh38
rs373499073 Chr.14: 35402468-35402468 on Build GRCh38
rs116801398 Chr.14: 35402396-35402396 on Build GRCh38
rs1016329748 Chr.14: 35402365-35402365 on Build GRCh38
rs758698302 Chr.14: 35402389-35402389 on Build GRCh38
rs757400212 Chr.14: 35402360-35402361 on Build GRCh38
rs778919047 Chr.14: 35402373-35402373 on Build GRCh38
rs759161424 Chr.14: 35402452-35402452 on Build GRCh38
rs989569540 Chr.14: 35402453-35402453 on Build GRCh38
rs983972574 Chr.14: 35402490-35402490 on Build GRCh38
rs201427068 Chr.14: 35402326-35402326 on Build GRCh38
rs374962052 Chr.14: 35402359-35402359 on Build GRCh38
rs149917774 Chr.14: 35402509-35402509 on Build GRCh38
rs747273949 Chr.14: 35402510-35402510 on Build GRCh38
rs751513869 Chr.14: 35402404-35402415 on Build GRCh38
rs1019323420 Chr.14: 35402378-35402378 on Build GRCh38
rs560842402 Chr.14: 35402331-35402331 on Build GRCh38
rs528542318 Chr.14: 35402522-35402522 on Build GRCh38
rs368801300 Chr.14: 35402374-35402374 on Build GRCh38
rs762379232 Chr.14: 35402426-35402426 on Build GRCh38

View Full Product Details