Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 68226601-68226848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R1
Entrez Gene ID
5295
Gene Name
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
UniGene
Hs.132225
Gene Chromosome Location
Chr.5: 68215737-68301821 on Build GRCh38
Transcripts
NM_181523.2 XM_005248542.3 XM_017009585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1728757 c.65T>C p.I22T Chr.5: 68226740-68226740 on Build GRCh38
6574491 c.93_94delTG p.V32fs*2 Chr.5: 68226768-68226769 on Build GRCh38
7429196 c.150T>C p.P50P Chr.5: 68226825-68226825 on Build GRCh38
7346055 c.43delA p.K16fs*15 Chr.5: 68226718-68226718 on Build GRCh38
3071832 c.37G>C p.D13H Chr.5: 68226712-68226712 on Build GRCh38
View More

View Full Product Details