Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_006218.3 | NP_006209.2 |
XM_006713658.3 | XP_006713721.1 | |
XM_011512894.2 | XP_011511196.1 | |
GenBank mRNA | AB210020.1 | - |
AI379480.1 | - | |
AK021510.1 | - | |
AK292940.1 | - | |
AK297203.1 | - | |
AK302608.1 | - | |
AU129890.2 | - | |
AW445128.1 | - | |
BC113601.1 | - | |
BC113603.1 | - | |
BQ228953.1 | - | |
BX437219.2 | - | |
BX473473.1 | - | |
BX484734.1 | - | |
DQ910907.1 | - | |
HM437241.1 | - | |
U79143.1 | - | |
Z29090.1 | - |