Gene Transcripts

Gene Symbol
PIK3CB
Entrez Gene ID
5291
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
Gene Aliases
P110BETA, PI3K, PI3KBETA, PIK3C1
Chromosome Location
Chr.3: 138652698 - 138834938 on Build GRCh38
Uni Gene
Hs.239818
Interrogated Sequence Translated Protein
RefSeq NM_001256045.1 NP_001242974.1
NM_006219.2 NP_006210.1
XM_005247530.2 XP_005247587.1
XM_006713659.3 XP_006713722.1
XM_011512895.2 XP_011511197.1
XM_011512896.1 XP_011511198.1
XM_017006619.1 XP_016862108.1
XM_017006620.1 XP_016862109.1
XM_017006621.1 XP_016862110.1
GenBank mRNA AI623206.1 -
AK293754.1 -
AK302937.1 -
BC003393.1 -
BC022049.2 -
BC033948.2 -
BC035432.2 -
BC045790.1 -
BC114432.1 -
CR749357.1 -
ENST00000477593.0 -
S67334.1 -

View Full Product Details