Gene Transcripts

Gene Symbol
JUN
Entrez Gene ID
3725
Gene Name
Jun proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, AP1, c-Jun
Chromosome Location
Chr.1: 58780791 - 58784113 on Build GRCh38
Uni Gene
Hs.696684
Interrogated Sequence Translated Protein
RefSeq NM_002228.3 NP_002219.1
GenBank mRNA BC002646.2 -
BC006175.1 -
BC009874.2 -
BC068522.1 -
BG831943.1 -
BI793062.1 -
BT019759.1 -
BU782554.1 -
CD370800.1 -
CR541724.1 -
CR541752.1 -
ENST00000371222.0 -

View Full Product Details