Gene Transcripts

Gene Symbol
MAPK1
Entrez Gene ID
5594
Gene Name
mitogen-activated protein kinase 1
Gene Aliases
ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK
Chromosome Location
Chr.22: 21759657 - 21867680 on Build GRCh38
Uni Gene
Hs.431850
Interrogated Sequence Translated Protein
RefSeq NM_002745.4 NP_002736.3
NM_138957.3 NP_620407.1
GenBank mRNA AI004470.1 -
AK055080.1 -
AK057143.1 -
AK090869.1 -
AK295188.1 -
AK307615.1 -
BC017832.1 -
BC099905.1 -
BQ774391.1 -
DQ399292.1 -
EF565108.1 -
ENST00000215832.0 -
Z11694.1 -
Z11695.1 -

View Full Product Details