Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001256045.1 | NP_001242974.1 |
NM_006219.2 | NP_006210.1 | |
XM_005247530.2 | XP_005247587.1 | |
XM_006713659.3 | XP_006713722.1 | |
XM_011512895.2 | XP_011511197.1 | |
XM_011512896.1 | XP_011511198.1 | |
XM_017006619.1 | XP_016862108.1 | |
XM_017006620.1 | XP_016862109.1 | |
XM_017006621.1 | XP_016862110.1 | |
GenBank mRNA | AI623206.1 | - |
AK293754.1 | - | |
AK302937.1 | - | |
BC003393.1 | - | |
BC022049.2 | - | |
BC033948.2 | - | |
BC035432.2 | - | |
BC045790.1 | - | |
BC114432.1 | - | |
CR749357.1 | - | |
ENST00000477593.0 | - | |
S67334.1 | - |