Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_010591.2 | NP_034721.1 |
GenBank mRNA | AK132019.1 | - |
AK133722.1 | - | |
AK133867.1 | - | |
AK135023.1 | - | |
AK140911.1 | - | |
AK159196.1 | - | |
AK159220.1 | - | |
AY423853.1 | - | |
BC002081.1 | - | |
BC021888.1 | - | |
BC032086.1 | - | |
BC055887.1 | - | |
BC094032.1 | - | |
BY106462.1 | - | |
J04115.1 | - | |
X12740.1 | - | |
X12761.1 | - |