Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001038663.1 | NP_001033752.1 |
NM_011949.3 | NP_036079.1 | |
GenBank mRNA | AK013699.1 | - |
AK018549.1 | - | |
AK035386.1 | - | |
AK043621.1 | - | |
AK048127.1 | - | |
AK048159.1 | - | |
AK087925.1 | - | |
AK132241.1 | - | |
AK148853.1 | - | |
AK163692.1 | - | |
AK193161.1 | - | |
AK193200.1 | - | |
AK205643.1 | - | |
AK209313.1 | - | |
AK210736.1 | - | |
AK211534.1 | - | |
AK216848.1 | - | |
AK218813.1 | - | |
BB648734.1 | - | |
BC006708.1 | - | |
BC058258.1 | - | |
D10939.1 | - | |
X58712.1 | - |