Gene Transcripts

Gene Symbol
Olr424
Entrez Gene ID
296686
Gene Name
olfactory receptor 424
Gene Aliases
Chromosome Location
Chr. 3: 21130500 - 21131492 on Build Rnor_6.0
UniGene ID
Rn.143017
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000392.1 NP_001000392.1 1 742

View Full Product Details