Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027179.1 | NP_081455.1 | 4 | 271 | |
NM_134118.5 | NP_598879.1 | 5 | 316 | ||
GenBank mRNA | AK002384.1 | - | 5 | 287 | |
AK010513.1 | - | 4 | 207 | ||
AK010984.1 | - | 5 | 290 | ||
AK013558.1 | - | 5 | 272 | ||
AK020708.1 | - | 4 | 265 | ||
BC019984.1 | - | 5 | 302 | 4240045 | |
BC094046.1 | - | 5 | 416 | 5064002 | |
BY278351.1 | - | 4 | 271 |