Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267623.1 | NP_001254552.1 | 5 | 545 | |
NM_001324236.1 | NP_001311165.1 | 6 | 671 | ||
NM_001324237.1 | NP_001311166.1 | 7 | 576 | ||
NM_001324238.1 | NP_001311167.1 | 6 | 579 | ||
NM_033453.3 | NP_258412.1 | 7 | 668 | ||
NM_181493.3 | NP_852470.1 | 7 | 617 | ||
NR_052000.1 | 6 | 511 | |||
NR_052001.2 | 6 | 589 | |||
NR_052002.1 | 6 | 603 | |||
XM_006723564.3 | XP_006723627.1 | 8 | 998 | ||
XM_006723565.3 | XP_006723628.1 | 6 | 875 | ||
GenBank mRNA | AB062127.1 | - | 7 | 537 | |
AF026816.2 | - | 7 | 527 | ||
AF219116.1 | - | 7 | 580 | ||
AF334680.1 | - | 6 | 460 | ||
BC010138.2 | - | 7 | 512 | 4124624 | |
BE275997.1 | - | 6 | 546 | ||
BF969192.1 | - | 6 | 511 | ||
BI115811.1 | - | 5 | 381 | ||
BM560812.1 | - | 668 | |||
BQ049090.1 | - | 6 | 450 | ||
EF199841.1 | - | 7 | 515 | ||
EF213026.1 | - | 7 | 464 | ||
KU178148.1 | - | 7 | 476 | ||
KU178149.1 | - | 5 | 353 |