Gene Transcripts
- Gene Symbol
- TP53I3
- Entrez Gene ID
- 9540
- Gene Name
- tumor protein p53 inducible protein 3
- Gene Aliases
- PIG3
- Chromosome Location
- Chr. 2: 24077428 - 24085215 on Build GRCh38
- UniGene ID
- Hs.50649
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004881.4 |
NP_004872.2
|
4, 5 |
1697 |
|
NM_147184.3 |
NP_671713.1
|
5, 6 |
1298 |
|
XM_005264650.2 |
XP_005264707.1
|
5, 6 |
1170 |
|
GenBank mRNA |
AF010309.1 |
- |
4, 5 |
1335 |
|
AK223382.1 |
- |
4, 5 |
1534 |
|
AK297579.1 |
- |
4, 5 |
1138 |
|
BC000474.2 |
- |
5, 6 |
868 |
2961571 |
BC018819.2 |
- |
5, 6 |
868 |
|
BT007149.1 |
- |
4, 5 |
808 |
|
CN285566.1 |
- |
1, 2 |
139 |
|
View Full Product Details