Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144996.1 | NP_001138468.1 | 2 | 525 | |
NM_002206.2 | NP_002197.2 | 2 | 525 | ||
XM_005268839.1 | XP_005268896.1 | 2 | 658 | ||
XM_005268840.1 | XP_005268897.1 | 2 | 658 | ||
XM_005268841.1 | XP_005268898.1 | 2 | 658 | ||
XM_005268842.1 | XP_005268899.1 | 2 | 658 | ||
XM_005268844.1 | XP_005268901.1 | 3 | 283 | ||
XM_005268846.1 | XP_005268903.1 | 3 | 196 | ||
GenBank mRNA | AB210036.1 | - | 2 | 495 | |
AF032108.1 | - | 2 | 466 | ||
AF052050.1 | - | 2 | 305 | ||
AF072132.1 | - | 2 | 305 | ||
AY358882.1 | - | 2 | 324 | ||
BC050280.1 | - | 2 | 521 | 5284443 |