Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031847.2 | NP_001027017.1 | 6 | 788 | |
NM_001876.3 | NP_001867.2 | 6 | 788 | ||
XM_005273762.2 | XP_005273819.1 | 6 | 764 | ||
XM_005273763.1 | XP_005273820.1 | 6 | 764 | ||
XM_017017220.1 | XP_016872709.1 | 6 | 897 | ||
GenBank mRNA | AK025307.1 | - | 1 | 74 | |
AK172798.1 | - | 6 | 774 | ||
AK309464.1 | - | 6 | 764 | ||
AK314301.1 | - | 6 | 727 | ||
BC000185.1 | - | 6 | 663 | 3352642 | |
BT009791.1 | - | 5 | 618 | ||
L39211.1 | - | 6 | 694 |