Gene Transcripts
- Gene Symbol
- PSKH2
- Entrez Gene ID
- 85481
- Gene Name
- protein serine kinase H2
- Gene Aliases
- Chromosome Location
- Chr. 8: 86047109 - 86069987 on Build GRCh38
- UniGene ID
- Hs.680136
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033126.1 |
NP_149117.1
|
1, 2 |
193 |
|
GenBank mRNA |
AY037806.1 |
- |
1, 2 |
193 |
|
View Full Product Details