Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000285.3 | NP_000276.2 | 11 | 930 | |
NM_001166056.1 | NP_001159528.1 | 9 | 807 | ||
NM_001166057.1 | NP_001159529.1 | 9 | 738 | ||
GenBank mRNA | AK225037.1 | - | 11 | 811 | |
AK290756.1 | - | 11 | 811 | ||
AK290781.1 | - | 11 | 826 | ||
AK291561.1 | - | 11 | 828 | ||
AK293126.1 | - | 9 | 633 | ||
AK294619.1 | - | 9 | 705 | ||
BC004305.1 | - | 11 | 801 | 3626134 | |
BC015027.1 | - | 11 | 831 | 3920275 | |
BC028295.1 | - | 11 | 802 | 4470066 | |
BT006692.1 | - | 11 | 797 | ||
CR541669.1 | - | 11 | 797 | ||
J04605.1 | - | 11 | 813 |