Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
Chromosome Location
Chr. 7: 112480219 - 112491062 on Build GRCh38
UniGene ID
Hs.396189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134468.1 NP_001127940.1 4 558
NM_182597.2 NP_872403.1 4 764
XM_011516074.1 XP_011514376.1 4 1267
XM_011516075.2 XP_011514377.1 4 2954
XM_011516076.2 XP_011514378.1 4 2590
XM_017012028.1 XP_016867517.1 3 2472
GenBank mRNA AK096894.1 - 4 780
BC126149.1 - 4 602 8991865
BC130568.1 - 4 602 40146599
DB456503.2 - 4 558

View Full Product Details