Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114171.1 | NP_001107643.1 | 3 | 3026 | |
NM_006732.2 | NP_006723.2 | 4 | 3134 | ||
XM_005258691.1 | XP_005258748.1 | 5 | 2993 | ||
GenBank mRNA | AF432216.1 | - | 1 | 1457 | |
BC036724.1 | - | 5 | 3112 | 5212854 | |
BC040197.1 | - | 3 | 3134 | 4816584 | |
BX538117.1 | - | 5 | 3277 | ||
EU178109.1 | - | 4 | 3026 | ||
EU178110.1 | - | 6 | 2993 | ||
EU178111.1 | - | 4 | 3017 | ||
EU178112.1 | - | 3 | 2909 | ||
EU178113.1 | - | 5 | 2885 | ||
EU178114.1 | - | 5 | 2876 | ||
EU178115.1 | - | 3 | 2705 | ||
EU178116.1 | - | 4 | 2564 | ||
L49169.1 | - | 5 | 3134 |