Gene Transcripts
- Gene Symbol
- ABCB8
- Entrez Gene ID
- 11194
- Gene Name
- ATP binding cassette subfamily B member 8
- Gene Aliases
- EST328128, M-ABC1, MABC1
- Chromosome Location
- Chr. 7: 151028422 - 151047782 on Build GRCh38
- UniGene ID
- Hs.647118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF047690.1 |
- |
5 |
775 |
|
View Full Product Details