Gene Transcripts
- Gene Symbol
- DNAH5
- Entrez Gene ID
- 1767
- Gene Name
- dynein axonemal heavy chain 5
- Gene Aliases
- CILD3, DNAHC5, HL1, KTGNR, PCD
- Chromosome Location
- Chr. 5: 13690328 - 14011829 on Build GRCh38
- UniGene ID
- Hs.212360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001369.2 |
NP_001360.1
|
3, 4 |
327 |
|
XM_005248262.3 |
XP_005248319.2
|
3, 4 |
410 |
|
XM_017009177.1 |
XP_016864666.1
|
3, 4 |
410 |
|
XM_017009178.1 |
XP_016864667.1
|
3, 4 |
1164 |
|
XM_017009180.1 |
XP_016864669.1
|
3, 4 |
410 |
|
XM_017009181.1 |
XP_016864670.1
|
3, 4 |
410 |
|
XM_017009182.1 |
XP_016864671.1
|
3, 4 |
410 |
|
XM_017009183.1 |
XP_016864672.1
|
3, 4 |
410 |
|
XM_017009184.1 |
XP_016864673.1
|
3, 4 |
410 |
|
XM_017009187.1 |
XP_016864676.1
|
3, 4 |
410 |
|
GenBank mRNA |
AK055138.1 |
- |
3, 4 |
643 |
|
AY045575.1 |
- |
3, 4 |
327 |
|
AY050643.1 |
- |
3, 4 |
329 |
|
View Full Product Details