Gene Transcripts

Gene Symbol
NAAA
Entrez Gene ID
27163
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
ASAHL, PLT
Chromosome Location
Chr. 4: 75910655 - 75941013 on Build GRCh38
UniGene ID
Hs.437365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005262925.3 XP_005262982.1 8, 9 1027
GenBank mRNA AK024677.1 - 1529
BC006388.2 - 7, 8 932 4122755

View Full Product Details