Gene Transcripts
- Gene Symbol
- FOLR2
- Entrez Gene ID
- 2350
- Gene Name
- folate receptor beta
- Gene Aliases
- BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3
- Chromosome Location
- Chr. 11: 72216775 - 72221950 on Build GRCh38
- UniGene ID
- Hs.433159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000803.4 |
NP_000794.3
|
2, 3 |
321 |
|
NM_001113534.1 |
NP_001107006.1
|
2, 3 |
315 |
|
NM_001113535.1 |
NP_001107007.1
|
2, 3 |
309 |
|
NM_001113536.1 |
NP_001107008.1
|
2, 3 |
303 |
|
XM_005273856.3 |
XP_005273913.1
|
2, 3 |
319 |
|
GenBank mRNA |
AK222539.1 |
- |
2, 3 |
312 |
|
AK222643.1 |
- |
2, 3 |
303 |
|
BC027894.1 |
- |
2, 3 |
290 |
|
BC058036.1 |
- |
2, 3 |
305 |
6662871 |
BC115366.1 |
- |
1, 2 |
91 |
|
BM691283.1 |
- |
2, 3 |
261 |
|
BM849995.1 |
- |
2, 3 |
321 |
|
BY797003.2 |
- |
2, 3 |
284 |
|
CD692073.1 |
- |
2, 3 |
319 |
|
View Full Product Details