Gene Transcripts

Gene Symbol
PAAF1
Entrez Gene ID
80227
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
PAAF, Rpn14, WDR71
Chromosome Location
Chr. 11: 73876699 - 73927736 on Build GRCh38
UniGene ID
Hs.525017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267803.1 NP_001254732.1 12 1174
NM_001267804.1 NP_001254733.1 12 1165
NM_001267805.1 NP_001254734.1 12 1161
NM_001267806.1 NP_001254735.1 11 1048
NM_025155.2 NP_079431.1 11 1105
XM_005274308.2 XP_005274365.2 9 1697
XM_005274309.1 XP_005274366.1 10 964
XM_017018357.1 XP_016873846.1 12 1093
XM_017018358.1 XP_016873847.1 12 1098
XM_017018359.1 XP_016873848.1 9 1656
GenBank mRNA AF087895.1 - 12 1099
AK021910.1 - 11 1086
AK222501.1 - 11 1086
AK225520.1 - 11 1073
AK298258.1 - 11 1048
AK309133.1 - 9 1312
AK316394.1 - 11 1319
BC006142.2 - 11 1058 3937356
BC021541.1 - 11 1069 3942680
BC028628.1 - 12 1139 5263875

View Full Product Details