Gene Transcripts

Gene Symbol
DLEU7
Entrez Gene ID
220107
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
Chromosome Location
Chr. 13: 50711026 - 50843939 on Build GRCh38
UniGene ID
Hs.673860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198989.3 NP_945340.2 2 985
GenBank mRNA AK126830.1 - 2 985
BC104892.1 - 2 795 8143895
BC112009.1 - 2 795 8327477

View Full Product Details