Gene Transcripts
- Gene Symbol
- SAMM50
- Entrez Gene ID
- 25813
- Gene Name
- SAMM50 sorting and assembly machinery component
- Gene Aliases
- CGI-51, OMP85, SAM50, TOB55, TRG-3, YNL026W
- Chromosome Location
- Chr. 22: 43955381 - 43996532 on Build GRCh38
- UniGene ID
- Hs.505824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015380.4 |
NP_056195.3
|
3, 4 |
430 |
|
GenBank mRNA |
AF151809.1 |
- |
3, 4 |
342 |
|
AK001087.1 |
- |
3, 4 |
369 |
|
AK222657.1 |
- |
3, 4 |
304 |
|
AK297770.1 |
- |
3, 4 |
265 |
|
AK312773.1 |
- |
3, 4 |
265 |
|
AY189688.1 |
- |
3, 4 |
345 |
|
BC007830.1 |
- |
3, 4 |
337 |
3635175 |
BC011681.2 |
- |
3, 4 |
390 |
3010765 |
BC015200.1 |
- |
3, 4 |
280 |
4333051 |
CR456483.1 |
- |
3, 4 |
242 |
|
DB030637.1 |
- |
3, 4 |
430 |
|
View Full Product Details