Gene Transcripts
- Gene Symbol
- SAMD3
- Entrez Gene ID
- 154075
- Gene Name
- sterile alpha motif domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 6: 130133203 - 130365425 on Build GRCh38
- UniGene ID
- Hs.440508
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001017373.3 |
NP_001017373.2
|
4, 5 |
595 |
|
NM_001258275.2 |
NP_001245204.1
|
6, 7 |
876 |
|
NM_001277185.1 |
NP_001264114.1
|
3, 4 |
377 |
|
XM_017010305.1 |
XP_016865794.1
|
3, 4 |
425 |
|
XM_017010306.1 |
XP_016865795.1
|
2, 3 |
635 |
|
XM_017010307.1 |
XP_016865796.1
|
2, 3 |
635 |
|
XM_017010308.1 |
XP_016865797.1
|
2, 3 |
288 |
|
XM_017010309.1 |
XP_016865798.1
|
2, 3 |
288 |
|
XM_017010310.1 |
XP_016865799.1
|
2, 3 |
288 |
|
GenBank mRNA |
AK091351.1 |
- |
6, 7 |
878 |
|
AK091882.1 |
- |
4, 5 |
595 |
|
AK302228.1 |
- |
3, 4 |
379 |
|
AK310075.1 |
- |
2, 3 |
631 |
|
BC029851.1 |
- |
2, 3 |
284 |
|
BC119757.1 |
- |
2, 3 |
284 |
|
BC119758.1 |
- |
2, 3 |
284 |
|
BC127653.1 |
- |
2, 3 |
282 |
|
BC127654.1 |
- |
2, 3 |
282 |
|
DA584636.1 |
- |
2, 3 |
567 |
|
View Full Product Details