Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152744.3 NP_689957.3 4 760
GenBank mRNA AY310398.1 - 4 760

View Full Product Details