Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017754.3 | NP_060224.3 | 5 | 612 | |
GenBank mRNA | AB126777.1 | - | 5 | 447 | |
BC137120.1 | - | 5 | 442 | 9021117 | |
BC137127.1 | - | 5 | 442 | 9021124 | |
BX640641.1 | - | 5 | 401 | ||
DB636320.1 | - | 5 | 612 |