Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr. 7: 100400826 - 100429239 on Build GRCh38
UniGene ID
Hs.105191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258008.1 NP_001244937.1 13, 14 1632
NM_017984.4 NP_060454.3 13, 14 1629
XM_005250479.2 XP_005250536.1 13, 14 2066
XM_006716035.3 XP_006716098.1 13, 14 2066
XM_006716036.2 XP_006716099.1 13, 14 1570
XM_006716037.3 XP_006716100.1 13, 14 2063
XM_006716038.3 XP_006716101.1 13, 14 2063
GenBank mRNA AK000919.1 - 8, 9 1016
AK093038.1 - 13, 14 1570
AK300745.1 - 13, 14 1488
AK302108.1 - 12, 13 1366
AL136735.1 - 13, 14 1500
BC002725.1 - 10, 11 1330 3631266
KU178677.1 - 8, 9 892
KU178678.1 - 8, 9 895
KU178679.1 - 8, 9 892
KU178680.1 - 8, 9 895
KU178681.1 - 8, 9 892

View Full Product Details