Gene Transcripts

Gene Symbol
CSRP3
Entrez Gene ID
8048
Gene Name
cysteine and glycine rich protein 3
Gene Aliases
CLP, CMD1M, CMH12, CRP3, LMO4, MLP
Chromosome Location
Chr. 11: 19182030 - 19210571 on Build GRCh38
UniGene ID
Hs.83577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003476.4 NP_003467.1 4 373
GenBank mRNA AF121260.1 - 3 263
AK313289.1 - 3 198
BC005900.2 - 2 163 4247505
BC024010.1 - 4 376 4826902
BC057221.1 - 3 196 4337632
U20324.1 - 3 179
U49837.1 - 3 191
U72899.1 - 3 179

View Full Product Details