Gene Transcripts

Gene Symbol
HRH4
Entrez Gene ID
59340
Gene Name
histamine receptor H4
Gene Aliases
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R
Chromosome Location
Chr. 18: 24460568 - 24487427 on Build GRCh38
UniGene ID
Hs.287388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143828.1 NP_001137300.1 2 2179
NM_001160166.1 NP_001153638.1 2 2279
NM_021624.3 NP_067637.2 3 2443
GenBank mRNA AF312230.1 - 3 2444

View Full Product Details