Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128142.1 | NP_001121614.1 | 3 | 934 | |
NM_001199682.1 | NP_001186611.1 | 4 | 986 | ||
NM_001278247.1 | NP_001265176.1 | 5 | 1119 | ||
NM_001278248.1 | NP_001265177.1 | 5 | 1166 | ||
NM_015485.4 | NP_056300.2 | 4 | 1033 | ||
NR_103483.1 | 4 | 1050 | |||
NR_103484.1 | 5 | 666 | |||
GenBank mRNA | AL050062.1 | - | 3 | 878 | |
BC010936.2 | - | 4 | 986 | ||
BX648581.1 | - | 1 | 4088 |