Gene Transcripts

Gene Symbol
SEC31B
Entrez Gene ID
25956
Gene Name
SEC31 homolog B, COPII coat complex component
Gene Aliases
SEC31B-1, SEC31L2
Chromosome Location
Chr. 10: 100486646 - 100519838 on Build GRCh38
UniGene ID
Hs.18889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015490.3 NP_056305.1 4605
GenBank mRNA AL080141.1 - 3713
BM719543.1 - 413

View Full Product Details