Gene Transcripts

Gene Symbol
LMAN2
Entrez Gene ID
10960
Gene Name
lectin, mannose binding 2
Gene Aliases
C5orf8, GP36B, VIP36
Chromosome Location
Chr. 5: 177331562 - 177351884 on Build GRCh38
UniGene ID
Hs.75864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006816.2 NP_006807.1 2 532
GenBank mRNA AK222609.1 - 2 299
AK292099.1 - 2 309
AK301607.1 - 3 216
AK308027.1 - 2 443
BC017263.2 - 2 312 4135210
DC404202.1 - 2 532
U10362.1 - 2 295

View Full Product Details