Gene Transcripts

Gene Symbol
WBP2
Entrez Gene ID
23558
Gene Name
WW domain binding protein 2
Gene Aliases
GRAMD6, WBP-2
Chromosome Location
Chr. 17: 75845699 - 75855420 on Build GRCh38
UniGene ID
Hs.514489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012478.3 NP_036610.2 8 894
XM_011524580.2 XP_011522882.1 7 680
GenBank mRNA AF087866.1 - 8 798
AK057881.1 - 8 799
AK094458.1 - 2 1020
AK128407.1 - 4 2712
AK293214.1 - 9 948
AL049981.1 - 2 74
BC007452.1 - 8 797 3140484
BC010616.1 - 8 807 4177319

View Full Product Details