Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042548.1 | NP_001036013.1 | 5 | 778 | |
NM_005102.2 | NP_005093.2 | 5 | 778 | ||
XM_006712153.2 | XP_006712216.1 | 5 | 600 | ||
GenBank mRNA | AB076184.1 | - | 5 | 770 | |
AB209719.1 | - | 5 | 577 | ||
AF113124.1 | - | 5 | 677 | ||
AK025274.1 | - | 4 | 458 | ||
BC018032.1 | - | 5 | 757 | ||
BC037566.1 | - | 5 | 743 | ||
BC047020.1 | - | 5 | 729 | ||
BC054043.1 | - | 5 | 712 | ||
BC064484.1 | - | 5 | 692 | ||
BC089390.1 | - | 5 | 724 | ||
BC110386.1 | - | 5 | 733 | 5183735 | |
BX537785.1 | - | 6 | 704 | ||
U60061.1 | - | 2 | 131 | ||
U69140.1 | - | 4 | 419 |