Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000638.3 | NP_000629.3 | 7 | 1386 | |
GenBank mRNA | AK298441.1 | - | 6 | 1435 | |
AK312968.1 | - | 7 | 1338 | ||
BC005046.1 | - | 7 | 1317 | 4040317 | |
X03168.1 | - | 7 | 1298 |