Gene Transcripts

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
Chromosome Location
Chr. 2: 73385758 - 73609919 on Build GRCh38
UniGene ID
Hs.184720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015120.4 NP_055935.4 3 698
GenBank mRNA AJ417593.2 - 3 698
AX751581.1 - 2 572

View Full Product Details