Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017956.2 | NP_001017956.1 | 6 | 974 | |
NM_001017957.2 | NP_001017957.1 | 6 | 974 | ||
NM_001017958.2 | NP_001017958.1 | 6 | 974 | ||
NM_001261420.1 | NP_001248349.1 | 6 | 974 | ||
NM_001261421.1 | NP_001248350.1 | 5 | 875 | ||
NM_001261422.1 | NP_001248351.1 | 5 | 818 | ||
NM_001261423.1 | NP_001248352.1 | 5 | 797 | ||
NM_006812.3 | NP_006803.1 | 6 | 974 | ||
XM_005268581.1 | XP_005268638.1 | 6 | 906 | ||
XM_006719200.1 | XP_006719263.1 | 6 | 906 | ||
XM_006719201.1 | XP_006719264.1 | 6 | 906 | ||
GenBank mRNA | AB002806.1 | - | 6 | 808 | |
AK129781.1 | - | 6 | 786 | ||
AK130733.1 | - | 6 | 784 | ||
AK291374.1 | - | 6 | 856 | ||
AK293435.1 | - | 5 | 609 | ||
AK294870.1 | - | 6 | 724 | ||
AK296770.1 | - | 6 | 784 | ||
AK298532.1 | - | 5 | 685 | ||
AK303291.1 | - | 6 | 942 | ||
AK303858.1 | - | 5 | 628 | ||
AL137691.1 | - | 3 | 948 | ||
BC000532.2 | - | 6 | 779 | 2964645 | |
BC007254.1 | - | 6 | 779 | 3027883 | |
BC023513.2 | - | 6 | 769 | 4123782 | |
CR456791.1 | - | 6 | 767 | ||
U41635.1 | - | 6 | 852 |