Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301036.1 | NP_001287965.1 | 4 | 1870 | |
NM_020698.3 | NP_065749.2 | 4 | 1851 | ||
GenBank mRNA | AB032971.1 | - | 4 | 1780 | |
AB073655.1 | - | 1 | 346 | ||
BC040535.1 | - | 4 | 1837 | 5264859 |