Gene Transcripts

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
Chromosome Location
Chr. 12: 110614027 - 110649130 on Build GRCh38
UniGene ID
Hs.211511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082537.2 NP_001076006.1 3 570
NM_001082538.2 NP_001076007.1 3 570
NM_001173975.2 NP_001167446.1 3 631
NM_001173976.1 NP_001167447.1 4 646
NM_001319680.1 NP_001306609.1 3 570
NM_001319681.1 NP_001306610.1 3 570
NM_001319682.1 NP_001306611.1 3 631
NM_024549.5 NP_078825.2 3 570
NR_135088.1 3 631
XM_005253934.3 XP_005253991.1 3 474
XM_005253935.3 XP_005253992.1 3 474
XM_006719594.2 XP_006719657.1 3 535
XM_006719595.2 XP_006719658.1 4 641
XM_006719596.2 XP_006719659.1 3 491
XM_006719597.3 XP_006719660.1 2 361
XM_006719598.2 XP_006719661.1 3 491
XM_006719599.2 XP_006719662.1 3 469
XM_006719600.2 XP_006719663.1 2 408
XM_011538733.2 XP_011537035.1 3 474
XM_011538734.2 XP_011537036.1 2 429
XM_011538735.1 XP_011537037.1 3 474
XM_011538737.2 XP_011537039.1 3 474
XM_011538738.2 XP_011537040.1 3 474
XM_017019964.1 XP_016875453.1 3 535
XM_017019965.1 XP_016875454.1 3 458
XM_017019966.1 XP_016875455.1 3 505
XM_017019967.1 XP_016875456.1 3 335
XM_017019968.1 XP_016875457.1 2 424
XM_017019969.1 XP_016875458.1 3 469
GenBank mRNA AK024780.1 - 4 544
AK055891.1 - 3 484
AK092775.1 - 3 532
AK295514.1 - 3 570
AK301732.1 - 3 435
AY358184.1 - 3 433
BC033811.1 - 3 517
BC040113.1 - 3 460 6054789
BC044885.1 - 3 493
BC062611.1 - 3 421 5184202
DN997186.1 - 3 530
DQ278868.1 - 3 495

View Full Product Details