Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136019.2 | NP_001129491.1 | 3 | 769 | |
NM_004107.4 | NP_004098.1 | 2 | 505 | ||
XM_005258657.1 | XP_005258714.1 | 2 | 324 | ||
XM_011526627.2 | XP_011524929.1 | 2 | 462 | ||
GenBank mRNA | AK074734.1 | - | 1 | 896 | |
AK075510.1 | - | 2 | 316 | ||
AK075532.1 | - | 3 | 419 | ||
AK299124.1 | - | 2 | 289 | ||
BC008734.2 | - | 2 | 449 | 3163446 | |
BC020421.1 | - | 1 | 702 | ||
BT006828.1 | - | 2 | 283 | ||
BX647163.1 | - | 3 | 389 | ||
DC312637.1 | - | 2 | 506 | ||
U12255.1 | - | 2 | 408 |