Gene Transcripts

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
Chromosome Location
Chr. 9: 128255829 - 128276007 on Build GRCh38
UniGene ID
Hs.155827
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004486.4 NP_004477.3 3 259
XM_005251930.1 XP_005251987.1 4 358
XM_005251931.1 XP_005251988.1 4 358
XM_005251932.1 XP_005251989.1 3 270
GenBank mRNA AF248953.1 - 3 210
BC069268.1 - 3 259 6340972

View Full Product Details