Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297765.1 | NP_001284694.1 | 4 | 506 | |
NM_004365.3 | NP_004356.2 | 4 | 506 | ||
GenBank mRNA | BC005383.1 | - | 4 | 476 | |
BC093793.1 | - | 4 | 442 | 7939638 | |
BC112040.1 | - | 4 | 442 | 8327508 | |
BM553126.1 | - | 4 | 486 | ||
BT006688.1 | - | 4 | 381 | ||
CB988525.1 | - | 4 | 528 | ||
CB998764.1 | - | 4 | 509 | ||
Y12473.1 | - | 5 | 381 |