Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270420.1 | NP_001257349.1 | 3 | 412 | |
NM_001270421.1 | NP_001257350.1 | 3 | 412 | ||
NM_032545.3 | NP_115934.1 | 3 | 412 | ||
XM_011511486.2 | XP_011509788.1 | 3 | 407 | ||
GenBank mRNA | AF312769.1 | - | 3 | 170 | |
AK315326.1 | - | 3 | 412 | ||
BC069508.1 | - | 3 | 194 | 7262189 | |
BC074825.2 | - | 3 | 173 | 30915323 | |
BC074826.2 | - | 3 | 173 | 30915540 | |
BC110080.1 | - | 3 | 285 | 40030641 | |
BC146897.1 | - | 3 | 289 | 9056825 | |
BM509218.1 | - | 3 | 346 | ||
CA777249.1 | - | 3 | 385 | ||
CA941621.1 | - | 3 | 344 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079530.1 | NP_001072998.1 | 3 | 515 | |
XM_005263737.4 | XP_005263794.1 | 3 | 397 | ||
GenBank mRNA | BQ777283.1 | - | 3 | 357 | |
BU784123.1 | - | 3 | 370 |