Gene Transcripts

Gene Symbol
CLEC9A
Entrez Gene ID
283420
Gene Name
C-type lectin domain family 9 member A
Gene Aliases
CD370, DNGR-1, DNGR1, UNQ9341
Chromosome Location
Chr. 12: 10030677 - 10066030 on Build GRCh38
UniGene ID
Hs.531189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207345.3 NP_997228.1 7 1050
GenBank mRNA AY358265.1 - 7 1050
EU339276.1 - 4 437

View Full Product Details