Gene Transcripts

Gene Symbol
SLC2A7
Entrez Gene ID
155184
Gene Name
solute carrier family 2 member 7
Gene Aliases
GLUT7
Chromosome Location
Chr. 1: 8992534 - 9026384 on Build GRCh38
UniGene ID
Hs.531239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207420.2 NP_997303.2 5 527
XM_011540824.2 XP_011539126.1 5 566
XM_011540825.2 XP_011539127.1 5 566
GenBank mRNA AY571960.2 - 5 527

View Full Product Details