Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001045558.1 | NP_001039023.1 | 8 | 760 | |
NM_008837.3 | NP_032863.2 | 8 | 819 | ||
GenBank mRNA | AK050224.1 | - | 7 | 664 | |
AK090155.1 | - | 752 | |||
BC023088.1 | - | 8 | 726 | ||
BC030363.1 | - | 1 | 439 | ||
BC048788.1 | - | 8 | 705 | 6488048 | |
BU057774.1 | - | 8 | 722 | ||
CA468666.1 | - | 8 | 640 | ||
CD363874.1 | - | 5 | 249 | ||
Z46720.1 | - | 7 | 508 |