Gene Transcripts
- Gene Symbol
- Sos1
- Entrez Gene ID
- 20662
- Gene Name
- son of sevenless homolog 1 (Drosophila)
- Gene Aliases
- 4430401P03Rik, 9630010N06, AI449023
- Chromosome Location
- Chr. 17: 80393752 - 80480453 on Build GRCm38
- UniGene ID
- Mm.360004
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009231.2 |
NP_033257.2
|
10 |
2287 |
|
GenBank mRNA |
AK140310.1 |
- |
10 |
2287 |
|
View Full Product Details